21147
AATB21147EA
537
EUR
InStock
AATB21147
Cal-520N™ potassium salt
Cal-520N™ potassium salt
Cal-520® provides a robust homogeneous fluorescence-based assay tool for detecting intracellular calcium mobilization.
- Improved background/signal ratio
- Suitable for all major fluorescence platforms
- Cell impermeant
Attention: For research use only.