21204
AATB21204EA
152
EUR
InStock
AATB21204
PDMPO, RatioWorks™
PDMPO
The existing pH probes are ill-adapted to study acidic organelles such as lysosomes, endosomes, spermatozoa and acrosomes because their fluorescence is significantly reduced at lower pH.
- Large stokes shift
- Bright signal
- High photostability
Attention: For research use only.