21208
AATB21208EA
460
EUR
InStock
AATB21208
Red 600 SE, Protonex™
Red 600 SE
Protonex™ red dye demonstrated pH-dependent fluorescence.
- pH-dependent fluorescence
- Multiple applications
- Suitable for all major fluorescence platforms
- Enhanced signal at high pH
Attention: For research use only.