21211
AATB21211EA
307
EUR
InStock
AATB21211
PDMPO Dextran, RatioWorks™
PDMPO Dextran
The existing pH probes are ill-adapted to study acidic organelles such as lysosomes, endosomes, phagosomes, spermatozoa and acrosomes because their fluorescence is significantly reduced at lower pH.
- pH-dependent fluorescence
- Multiple applications
- Suitable for all major fluorescence platforms
- Improved cell retention
- Ratiometric signal
Attention: For research use only.