Amplite® Fluorimetric Sodium Ion Quantification Kit
Fournisseur: AAT BIOQUEST INC
21325
AATB21325EA 660 EUR InStock
AATB21325
Amplite® Fluorimetric Sodium Ion Quantification Kit
Tests Tests cellulaires
Designed for consistent and dependable results. Amplite® fluorimetric sodium ion quantification kit uses our robust sodium ion indicator dye, SoNa™ 520, which exhibit great fluorescence intensity enhancement upon binding to sodium ions.
  • Robust protocol
  • SoNa™ 520 is perhaps the most robust sodium ion indicator with high selectivity
  • 12 month shelf life
This microplate-based assay kit requires extremely small amount of sample, it is particularly suitable for the determination of sodium ion concentration in microvolume format.
Information de livraison: Shipped at RT.
Attention: Research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité