Portelite™ Fluorimetric Sodium Ion Quantification Kit
Fournisseur: AAT BIOQUEST
21327
AATB21327EA 660 EUR InStock
AATB21327
Portelite™ Fluorimetric Sodium Ion Quantification Kit
Tests Tests cellulaires
Designed for consistent and dependable results. Portelite™ fluorimetric sodium ion quantification kit uses our robust sodium ion indicator dye, SoNa™ 520, which exhibit great fluorescence intensity enhancement upon binding to sodium ions.
  • Robust protocol
  • This nanodrop-based assay kit requires a small amount of sample, it is particularly suitable for the determination of sodium ion concentration in fields or on site
  • 12-month shelf life
SoNa™ 520 is perhaps the best sodium ion indicator for rapidly determining sodium ion concentration in combination with a fluorescence device such as a fluorescence Nanodrop spectrophotometer or a fluorescence microplate reader.
Information de livraison: Shipped at RT.
Attention: Research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité