Amplite® Fluorimetric Lithium Ion Quantification Kit
Fournisseur: AAT BIOQUEST INC
21351
AATB21351EA 660 EUR InStock
AATB21351
Amplite® Fluorimetric Lithium Ion Quantification Kit
Tests Tests cellulaires
Designed for consistent and dependable results. Amplite® fluorimetric lithium ion quantification kit uses our new robust lithium-ion indicator dye, Lithiumighty™ 520, which exhibits great fluorescence intensity enhancement upon binding to lithium ions.
  • Lithiumighty ™ 520 is perhaps the most robust lithium-ion indicator with high selectivity
  • Requires very small amount of sample
  • 12 month shelf life
Microplate-based assay kit is particularly suitable for the determination of lithium ion concentration in a microvolume format.
Information de livraison: Shipped at RT.
Attention: Research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité