Portelite™ Fluorimetric Lithium Ion Quantification Kit
Fournisseur: AAT BIOQUEST
21353
AATB21353EA 687 EUR InStock
AATB21353
Portelite™ Fluorimetric Lithium Ion Quantification Kit
Tests Tests cellulaires
Designed for consistent and dependable results. Portelite™ fluorimetric lithium ion quantification kit uses our robust lithium ion indicator dye, Lithiumighty™ 520, which exhibit great fluorescence intensity enhancement upon binding to lithium ions.
  • Lithiumighty ™ 520 is perhaps the most robust lithium-ion indicator with high selectivity
  • Rapid determination of lithium concentrations in a variety of samples
  • 12-month shelf life
This nanodrop-based assay kit requires a small amount of sample, it is particularly suitable for the determination of lithium ion concentration in fields or on site.
Information de livraison: Shipped at RT.
Attention: Research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité