21411
AATB21411EA
146
EUR
InStock
AATB21411
DiBAC₄(3) pour la mesure du potentiel membranaire
DiBAC₄(3)
DiBAC4(3) is a sensitive slow-response probe for measuring cellular membrane potential.
- Sensitive detection
- Slow response probe
- Multiple applications