21414
AATB21414EA
146
EUR
InStock
AATB21414
DiSBAC2(3) pour la mesure du potentiel membranaire
DiSBAC2(3)
DiSBAC2(3) is a sensitive slow-response membrane potential probe that is widely used for measuring membrane potentials of many biological systems.
- Sensitive detection
- Slow response probe
- Multiple applications