PhosphoWorks™ Fluorimetric Pyrophosphate Assay Kit, Enhanced Selectivity
Fournisseur: AAT BIOQUEST
21614
AATB21614EA 537 EUR InStock
AATB21614
PhosphoWorks™ Fluorimetric Pyrophosphate Assay Kit, Enhanced Selectivity
Tests Tests cellulaires
Pyrophosphate (PPi) is produced by a number of biochemical reactions, such as ATP hydrolysis, DNA and RNA polymerizations, cyclic AMP formation by the enzyme adenylate cyclase and the enzymatic activation of fatty acids to form their coenzyme A esters.
  • Pyrophosphate-dependent fluorescence
  • Highly selective for PPi
  • Contains all necessary components
Attention: For research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité