Synonyms:
TF1 CPG
2240
AATB2240EA
307
EUR
InStock
AATB2240
Tide Fluor™ 1 CPG 500 Å, Tide Fluor™ sonde fluorescente
Tide Fluor™ 1 CPG
TF1 is designed to be a superior fluorophore alternative to EDANS.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity