Live or Dead™ Fixable Dead Cell Staining Kit
Fournisseur: AAT BIOQUEST
22598
AATB22598EA 266 EUR InStock
AATB22598
Live or Dead™ Fixable Dead Cell Staining Kit
Tests Tests cellulaires
IR Fluorescence. Our Live or Dead™ fixable dead cell staining kits are a set of tools for labelling cells for fluorescence microscopic investigations of cellular functions.
  • The kit provides all the essential components with an optimized cell-labelling protocol
  • The fluorescent dye used in the kit is well excited with the IR laser (808 nm) and emits fluorescence at 878 nm (885/40 bandpass filter).
This particular kit is designed to uniformly label fixed mammalian cells in infrared fluorescence for flow cytometry applications with IR laser excitation.
Information de livraison: Shipped at RT.
Attention: Research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité