2503
AATB2503EA
152
EUR
InStock
AATB2503
Cross linked-AlloPhycocyanin, ReadiUse™ sonde fluorescente
Cross linked-AlloPhycocyanin
Cross linked-Allophycocyanin (CL-APC) is a phycobiliprotein isolated from Spirulina sp., a blue-green alga.
- No dialysis required
- Ready to use
- High purity