2625
AATB2625EA
307
EUR
InStock
AATB2625
APC-Cy7 Tandem sonde fluorescente
APC-Cy7 Tandem
Allophycocyanin (APC) is a phycobiliprotein isolated from Spirulina sp., a blue-green alga.
- Suitable for most fluorescence platforms
- Excellent signal
- High quality