Synonyms:
BNTA
3008
AATB3008EA
137
EUR
InStock
AATB3008
Biotin NTA biodetection
Biotin NTA
Biotin NTA (BNTA), a biotin-labeled nitrilotriacetic acid, is widely used to detect histidine-tagged proteins immobilized on nitrocellulose membranes with the detection limite of ~0.1 pmol.