Native Human arginine-glycine-aspartic acid (from Human), iFluor® 750
Fournisseur: AAT BIOQUEST
36800
AATB36800EA 442 EUR InStock
AATB36800
Native Human arginine-glycine-aspartic acid (from Human), iFluor® 750
Protéines et Peptides
Near-infrared fluorescence optical imaging (also called in vivo Imaging) is emerging as a powerful technology for studying diseases at molecular level in small animal models.
  • Nir emission (757/779 nm)
  • Brighter and more stable signal
  • Multiple research applications
  • Suitable for both in vivo and in vitro
Attention: For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité