Amplite® Colorimetric Glucose and Sucrose Assay Kit
40009
AATB40009
Amplite® Colorimetric Glucose and Sucrose Assay Kit
Tests Tests enzymatiques
Designed for consistent and dependable results. The Amplite® Colorimetric Glucose and Sucrose Assay Kit offers a convenient method for measuring glucose and sucrose levels in various biological samples, such as serum, plasma, body fluids, food, tissue extracts, and growth medium.
  • Useful for assessing aerobic glycolysis, pyruvate kinase activity, and metabolic profiles in different biological matrices.
  • Measured at 570 nm
It is also employed for determining protein and sugar concentrations in food samples, as well as monitoring plasma insulin and glucose levels in murine models.
Information de livraison: Shipped with blue ice.
Attention: Research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité