Amplite® Fluorimetric Glucose and Sucrose Assay Kit
Fournisseur: AAT BIOQUEST INC
40010
AATB40010EA 647 EUR InStock
AATB40010
Amplite® Fluorimetric Glucose and Sucrose Assay Kit
Tests Tests enzymatiques
Designed for consistent and dependable results. The Amplite® fluorimetric glucose and sucrose assay kit offers a convenient method for measuring glucose and sucrose levels in various biological samples, such as serum, plasma, body fluids, food, tissue extracts, and growth medium.
  • Useful for assessing aerobic glycolysis, pyruvate kinase activity, and metabolic profiles in different biological matrices.
  • Ex/Em = 571/584 nm
It is also employed for determining protein and sugar concentrations in food samples, as well as monitoring plasma insulin and glucose levels in murine models.
Information de livraison: Shipped with blue ice.
Attention: Research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité