iFluor® 440, Styramide *Superior Replacement for Alexa Fluor 440 tyramide*
Fournisseur: AAT BIOQUEST INC
44900
AATB44900EA 413 EUR InStock
AATB44900
iFluor® 440, Styramide *Superior Replacement for Alexa Fluor 440 tyramide*
iFluor®
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10 to 50 times higher than the widely used tyramide (TSA) reagents.
In combination with our superior iFluor® dyes that have higher florescence intensity, increased photostability and enhanced water solubility, the iFluor® dye-labeled Styramide™ conjugates can generate fluorescence signal with significantly higher precision and sensitivity (more than 100 times) than standard ICC/IF/IHC. PSA utilises the catalytic activity of horseradish peroxidase (HRP) for covalent deposition of fluorophores in situ. PSA radicals have much higher reactivity than tyramide radicals, making the PSA system much faster, more robust and sensitive than the traditional TSA reagents.
Compared to tyramide reagents, the Styramide™ conjugates have ability to label the target at higher efficiency and thus generate significantly higher fluorescence signal. Styramide™ conjugates also allow significantly less consumption of primary antibody compared to standard directly conjugate method or tyramide amplification with the same level of sensitivity. iFluor® 440 Styramide is a new unique PSA reagent for multicolor application with our existing PSA and TSA reagents. AAT Bioquest offers the largest collection of TSA regents. We are the exclusive source of the superior PSA reagents for multicolor applications.
Attention: For research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité