4503
AATB4503EA
295
EUR
InStock
AATB4503
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) pour synthèse peptidique
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)
Compared to SMCC and sulfo-SMCC, SMCC Plus has enhanced water solubility and crosslinking efficiency.
- Improved crosslinking efficiency
- High water solubility