PSA™ iFluor® 555 Imaging Kit with Goat Anti-Rabbit IgG
Fournisseur: AAT BIOQUEST INC
45220
AATB45220EA 739 EUR InStock
AATB45220
PSA™ iFluor® 555 Imaging Kit with Goat Anti-Rabbit IgG
Systèmes de liaison et de marquage des protéines
iFluor® 555 PSA kit is a much superior replacement for Alexa Fluor 555 tyramide-based kit or other spectrally similar fluorescent tyramide or TSA kits.
  • Higher efficiency
  • Improved fluorescence signal
  • Less consumption of primary antibody
  • 10 to 50X improvement over TSA reagents
In combination with our superior iFluor® dyes that have higher florescence intensity, increased photostability and enhanced water solubility, the iFluor® dye-labeled Styramide™ conjugates can generate fluorescence signal with significantly higher precision and sensitivity (more than 100 times) than standard ICC/IF/IHC.
Conditionnement: Plastic vial.
Attention: For research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité