LRB Red™ Succinimidyl ester
Fournisseur: AAT BIOQUEST INC
471
AATB471EA 221 EUR InStock
AATB471
LRB Red™ Succinimidyl ester
Tests Tests cellulaires
Although Lissamine Rhodamine B chloride is one of the most popular labeling reagents of sulfonyl chloride, it is quite unstable in water. Even in anhydrous DMF, Lissamine Rhodamine B chloride tends to give a very complicated reaction mixture. It reacts with thiols, alcohols, phenols, aliphatic amines and aromatic amines indiscriminatingly. LRB Red™ is a succinimidyl ester and has the spectral properties identical to those of Lissamine Rhodamine B chloride. LRB Red™ is a superior replacement for Lissamine Rhodamine B chloride. LRB Red™ only reacts with aliphatic amines such as amino acids, peptides and proteins to give bright red fluorescent conjugates that are extremely stable. Compared to Lissamine Rhodamine B chloride, LRB Red™ has much higher labeling efficiency, and more importantly the resulted conjugates are much easier to be purified due to its much cleaner reactions. We strongly recommend that you consider using LRB Red™ to replace Lissamine Rhodamine B chloride for labeling peptides and oligonucleotides.
  • Identical spectra to Lissamine Rhodamine B chloride
  • High Labeling Efficiency
Attention: For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité