5020
AATB5020EA
146
EUR
InStock
AATB5020
Fmoc-Lys(biotin)-OH pour le marquage biomoléculaire
Fmoc-Lys(biotin)-OH
FMOC-Lys(Biotin)-OH is a lysine derivative for the preparation of biotin-labeled peptides by FMOC-SPPS.
- FRET building block
- Multiple applications