Amplite® Rapid Colorimetric APC-Maleimide Quantitation Kit
Fournisseur: AAT BIOQUEST INC
5527
AATB5527EA 297 EUR InStock
AATB5527
Amplite® Rapid Colorimetric APC-Maleimide Quantitation Kit
Tests Tests enzymatiques
Designed for consistent and dependable results. The Amplite® Rapid Colorimetric APC-Maleimide Quantitation Kit facilitates the quantification of maleimide groups on maleimide-activated APC proteins using a proprietary sensor, Maleimide 555™, which has an absorption peak at ~555 nm.
  • Robust protocol
  • This quantitation kit is compatible with various detection systems, including traditional cuvettes, NanoDrop™ Spectrophotometers, and 96-well absorbance plate readers
  • Six-month shelf life
Maleimide-based crosslinking reagents are commonly used for conjugating proteins to other proteins or biomolecules.
Information de livraison: Shipped with blue ice.
Attention: Research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité