Beadlite™ Rapid Colorimetric Thiol Quantitation Kit for Nanoparticles
Fournisseur: AAT BIOQUEST INC
5535
AATB5535EA 386 EUR InStock
AATB5535
Beadlite™ Rapid Colorimetric Thiol Quantitation Kit for Nanoparticles
Tests Tests enzymatiques
Designed for consistent and dependable results. Beadlite™ Rapid Colorimetric Thiol Quantitation kit provides a robust method to quantify thiol-functionalised nanoparticle with our proprietary Thiolite™ 490 that has its maximum absorbance at ~490 nm.
  • Robust protocol
  • Can be performed in a traditional cuvette, NanoDrop™ Spectrophotometer or a convenient 96-well absorbance plate reader
  • 12-month shelf life
This kit has been widely used for the rapid quantification of sulfhydryl groups on the surface of latex beads, agarose beads, or magnetic nanoparticles.
Information de livraison: Shipped with blue ice.
Attention: Research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité