5605
AATB5605EA
307
EUR
InStock
AATB5605
cOVA antibody development
cOVA
cOVA is the ovalbumin (OVA) modified with ethylenediamine to make the proteins bear more amine groups (thus more positive charges at neutral pH).
- Protein modifier
- Increased immunological response