5606
AATB5606EA
307
EUR
InStock
AATB5606
hOVA antibody development
hOVA
hOVA is the ovalbumin (OVA) modified with hexamethylenediamine to make the proteins bear more amine groups (thus more positive charges at neutral pH).
- Protein modifier
- Increased immunological response