Synthetic Maleimide Activated mKLH (from Megathura crenulata)
Fournisseur: AAT BIOQUEST INC
5612
AATB5612EA 517 EUR InStock
AATB5612
Synthetic Maleimide Activated mKLH (from Megathura crenulata)
Protéines et Peptides
Maleimide Activated mKLH is engineered to facilitate the conjugation of sulfhydryl-containing peptide haptens to the highly immunogenic keyhole limpet hemocyanin (KLH) carrier protein, optimising it for immunisation and antibody production.
  • Designed for heightened immunogen response
This high-purity, mariculture-derived KLH is chemically modified with a Sulfo-SMCC crosslinker, rendering it reactive to sulfhydryl groups (-SH) on cysteine residues of peptides and other thiol-containing haptens.
Attention: For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité