6001
AATB6001EA
739
EUR
InStock
AATB6001
Chemical Phosphorylation Reagent I (CPR I)
Chemical Phosphorylation Reagent I (CPR I)
This reagent can be used to create the 3'-phosphate by adding it as the first addition to the support.
- Oligonucleotide synthesis reagent
- Useful for bioconjugate development