ReadiPrep™ Lipopolysaccharide (LPS) Isolation Kit
Fournisseur: AAT BIOQUEST
60201
AATB60201EA 266 EUR InStock
AATB60201
ReadiPrep™ Lipopolysaccharide (LPS) Isolation Kit
Tests Tests cellulaires
Designed for consistent and dependable results. The ReadiPrep™ lipopolysaccharide (LPS) isolation kit provides a safe and effective way to isolate LPS from the outer membrane of Gram-negative bacteria.
  • Robust protocol
  • Unlike conventional approaches that rely on hazardous organic chemicals like chloroform or phenol, this kit provides a safer and more time-efficient alternative for isolating high-purity LPS for research applications
  • Six-month shelf life
Kit utilises a bacterial membrane lysis buffer and enzymatic protein digestion to extract pure LPS from bacterial cultures, which can be quantified using carbohydrate detection methods.
Information de livraison: Shipped with blue ice.
Attention: Research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité