DY547 CEP (DyLight 547 Phosphoramidite)
Fournisseur: AAT BIOQUEST INC
6074
AATB6074EA 669 EUR InStock
AATB6074
DY547 CEP (DyLight 547 Phosphoramidite)
Systèmes de liaison et de marquage des protéines
DY547 CEP (DyLight 547 phosphoramidite) is an excellent replacement to the commonly used Cy3 phosphoramidite.
  • High stability
  • Oligo deprotection can be carried out at room temperature
DY547 CEP appears to be more stable than Cy3 phosphoramidite. As Cy3 and Cy5 phosphoramidites, the deprotection of oligos containing DY547 CEP may be carried out with ammonium hydroxide at room temperature, regardless of the base protecting groups on the monomers used..
It is always prudent to use monomers with base labile protecting groups to limit the exposure time to 2 hours or less at 65 °C during deprotection.
Conditionnement: Plastic vial.
Attention: For research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité