DY647 CEP (DyLight 647 phosphoramidite)
Fournisseur: AAT BIOQUEST INC
6076
AATB6076EA 683 EUR InStock
AATB6076
DY647 CEP (DyLight 647 phosphoramidite)
Systèmes de liaison et de marquage des protéines
DY647 CEP (DyLight 647 phosphoramidite) is an excellent replacement to the commonly used Cy5 phosphoramidite.
  • High stability
  • Oligo deprotection can be carried out at room temperature
DY647 CEP appears to be more stable than Cy5 phosphoramidite. As Cy3 and Cy5 phosphoramidites, the deprotection of oligos containing DY647 CEP may be carried out with ammonium hydroxide at room temperature, regardless of the base protecting groups on the monomers used.
It is always prudent to use monomers with base labile protecting groups to limit the exposure time to 2 hours or less at 65 °C during deprotection.
Conditionnement: Plastic vial.
Attention: For research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité