DY677 CEP (DyLight 677 Phosphoramidite)
Fournisseur: AAT BIOQUEST
6078
AATB6078EA 977 EUR InStock
AATB6078
DY677 CEP (DyLight 677 Phosphoramidite)
Systèmes de liaison et de marquage des protéines
DY677 CEP (DyLight 677 phosphoramidite) is an excellent replacement to the commonly used Cy5.5 phosphoramidite.
  • High stability
  • Oligo deprotection can be carried out at room temperature
DY677 CEP appears to be more stable than Cy5.5 phosphoramidite. As Cy5.5 phosphoramidite, the deprotection of oligos containing DY677 CEP may be carried out with ammonium hydroxide at room temperature, regardless of the base protecting groups on the monomers used.
It is always prudent to use monomers with base labile protecting groups to limit the exposure time to 2 hours or less at 65 °C during deprotection.
Conditionnement: Plastic vial.
Attention: For research use only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité