ROX Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
Fournisseur: AAT BIOQUEST INC
67004
AATB67004EA 130 EUR InStock
AATB67004
ROX Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
Microplaques Plaques PCR
The ROX dye qPCR calibration plate can be used to maintain the 7500 real-time PCR system with fast 96-well block.
  • The qPCR calibration plate might significantly improve qPCR results with multiplexing by more accurately representing fluorescent spectra used in your real-time experiments
  • This calibration plate is ready to use without any additional preparation steps required
For most qPCR instruments, the necessary calibrations should be run at least every six months.
Attention: Avoid freeze-thawing plates. It is not recommended to use the plates more than 3 times. Plates can be stored for up to 12 months. Freeze (<−15 °C), and minimise light exposure.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité