9000
AATB9000EA
439
EUR
InStock
AATB9000
Fluorescein aldehyde [5-FAM aldehyde] pour le marquage biomoléculaire
Fluorescein aldehyde [5-FAM aldehyde]
Fluorescein aldehyde is a reactive fluorescent dye that can react with an amine, hydrazine or hydroxylamine.
- Reactive with amine, hydrazine, or hydroxylamine
- Detectable with the FITC channel