9010
AATB9010EA
439
EUR
InStock
AATB9010
Cy3 aldehyde pour le marquage biomoléculaire
Cy3 aldehyde
Cy3 aldehyde is a reactive fluorescent dye that can react with an amine, hydrazine or hydroxylamine.
- Reactive with amine, hydrazine, or hydroxylamine
- Detectable with the FITC channel