9011
AATB9011EA
422
EUR
InStock
AATB9011
Cy5 aldehyde pour le marquage biomoléculaire
Cy5 aldehyde
Cy5 aldehyde is a reactive red fluorescent dye that can react with an amine, hydrazine or hydroxylamine.
- Reactive with amine, hydrazine, or hydroxylamine
- Detectable with the FITC channel