Estrogen Receptor (ERα) (Human) ELISA Kit
Fournisseur: Abcam
AB285240-96
ABCAAB285240-96EA 800 EUR InStock
ABCAAB285240-96
Estrogen Receptor (ERα) (Human) ELISA Kit
Tests Tests ELISA
An Sandwich ELISA kit to quantify human ER-alpha in human serum, plasma and other biological fluids.
  • Easy, convenient and time-saving method to measure the level of ER-alpha in human serum, plasma, tissue homogenates, culture supernatants and other biological fluids
  • Detection Range: 62,5 to 4000 pg/ml
  • Sensitivity: <37,5 pg/ml
Estrogen Receptor is a nuclear hormone receptor. The steroid hormones and their receptors are involved in the regulation of eukaryotic gene expression and affect cellular proliferation and differentiation in target tissues. Ligand-dependent nuclear transactivation involves either direct homodimer binding to a palindromic estrogen response element (ERE) sequence or association with other DNA-binding transcription factors, such as AP-1/c-Jun, c-Fos, ATF-2, Sp1 and Sp3, to mediate ERE-independent signaling. Ligand binding induces a conformational change allowing subsequent or combinatorial association with multiprotein coactivator complexes through LXXLL motifs of their respective components. Mutual transrepression occurs between the estrogen receptor (ER) and NF-kappa-B in a cell-type specific manner. BioVision’s ER-alpha ELISA kit is a sandwich ELISA assay for the quantitative measurement of human ER-alpha in serum, plasma and cell culture supernatants. The density of colour is proportional to the amount of human ER-alpha captured from the samples.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité