Gentamicin (serum/urine) ELISA Kit
Fournisseur: Abcam
AB285241-96
ABCAAB285241-96EA 785 EUR InStock
ABCAAB285241-96
Gentamicin (serum/urine) ELISA Kit
Tests Tests ELISA
A competitive ELISA kit to quantify Gentamicin in Serum, urine, milk and tissues.
  • Easy, convenient and time-saving method to measure the level of Gentamicin in tissues, milk and liquid samples
  • Detection Range: 0,3 to 24,3 ppb (ng/ml)
  • Sensitivity: 0,3 ppb
  • Detection limitation: 3 ppb for tissues, milk and liquid samples
Gentamicin, an aminoglycosidic antibiotic, has a broad spectrum towards most bacteria, particularly gram-negative species. It is widely used by veterinaries, has industrials applications as an additive and can be found on foodstuff consumed by animals. Its popularity is due to its broad antibacterial spectrum, efficacy and low cost. It is also known that unregulated and excessive use of this antibiotic causes negative effects in humans (i.e. neurological, hearing and renal damage). Due to its dangerous side effects, the FDA has established strict parameters that are considered safe for human consumption. Consequently, monitoring Gentamicin concentration amounts in animal-derived food is critical. Standard techniques/instruments (HPLC or GC-MS) are utilized to detect Gentamicin. However, these techniques are complex, expensive, laborious, and time-consuming. Immunoassay techniques, such as ELISAs are commonly preferred as a simple, reliable and rapid method for the quantification of Gentamicin in various samples. BioVision’s Gentamicin ELISA Kit is a competitive-based ELISA that can be used for the determination of this antibiotic in dairy products, mammalian tissues and human biofluids. This Gentamicin detection kit is fast, can be completed within 90 min, offers ready-to-use reagents and can detect Gentamicin within a range between 0,3 to 24,3 ppb (ng/ml).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité