QuickDetect™ Total Cholesterol (Mouse) ELISA Kit
Fournisseur: Abcam
AB285242-96
ABCAAB285242-96EA 985 EUR InStock
ABCAAB285242-96
QuickDetect™ Total Cholesterol (Mouse) ELISA Kit
Tests Tests ELISA
Rapid sandwich ELISA kit to quantify Total cholesterol in mouse serum, plasma, culture supernatnat.
  • Easy, convenient and time-saving method to measure the level of total Cholesterol in mouse serum, plasma, culture supernatants and other biological fluids
  • The entire assay can be performed within 90 minutes with only a single incubation and a wash step respectively, resulting in fewer handling steps, fewer errors and more consistent results
  • Detection Range: 78 to 5000 pg/ml
Cholesterol, is an organic molecule, a sterol (or modified steroid), a type of lipid molecule, and is biosynthesised by all animal cells, in which it is an essential structural component of animal cell membranes that is required to maintain both membrane structural integrity and fluidity. Cholesterol enables animal cells to dispense with a cell wall, thereby allowing animal cells to change shape and animals to move. Total cholesterol is a measure of the total amount of cholesterol in blood, including low-density lipoprotein (LDL) cholesterol and high-density lipoprotein (HDL) cholesterol. BioVision’s QuickDetect™ total cholesterol ELISA kit is a sandwich ELISA assay for measuring the quantity of mouse total cholesterol in serum, plasma and cell culture and other biological fluid. The entire process can be performed within 90 minutes with only a single incubation and a wash step respectively, resulting in fewer handling steps, fewer errors and more consistent results.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité