Ciprofloxacin (Cipro) ELISA Kit
Fournisseur: Abcam
AB285266-96
ABCAAB285266-96EA 980 EUR InStock
ABCAAB285266-96
Ciprofloxacin (Cipro) ELISA Kit
Tests Tests ELISA
A competitive ELISA kit for quantify of Ciprofloxacin in tissue, honey, eggs, milk, milk powder.
  • Easy, convenient and time-saving method to measure the level of Ciprofloxacin in tissue, honey, eggs, milk, milk powder
  • Detection Range: 0,5 to 40 ppb (ng/ml)
  • Sensitivity: <0,05 ppb
  • Detection limit: 0,3 ppb for tissue, 0,4 ppb for honey, 3 ppb for milk and egg, 6 ppb for milk powder
  • Cross Reactivity: Sarafloxacin −110%; Oxolin acid −28%; Levofloxacin −10%; Lomefloxacin, Marbofloxacin −4%
Ciprofloxacin is an antibiotic used to treat a number of bacterial infections. It is commonly used to treat acne and rosacea. Historically it was important in reducing the number of deaths from cholera. A broad-spectrum antibiotic of the polyketide class, it is produced by the actinobacterial genus Streptomyces. It acts by inhibiting protein synthesis. It is first-line therapy for rocky mountain spotted fever, Lyme disease, Q fever, psittacosis and lymphogranuloma venereum, mycoplasma pneumoniae and to eradicate nasal carriage of meningococci. BioVision’s Ciprofloxacin ELISA kit is a competitive ELISA assay for the quantitative measurement of Ciprofloxacin in tissues, honey and urine. The density of colour is proportional to the amount of tetracycline captured from the samples.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité
Ce site utilise des cookies, en provenance de VWR ou de ses partenaires, afin de collecter des informations statistiques sur votre navigation et vous proposer des contenus en accord avec vos préférences, générés en fonction de vos habitudes de navigation. En poursuivant la consultation de ce site, vous approuvez l’utilisation de ces cookies.
Pour en savoir plus, consultez notre Politique de Confidentialit&eacute;..