Neomycin ELISA Kit, Inc.
Fournisseur: Abcam
AB285276-96
ABCAAB285276-96EA 1065 EUR InStock
ABCAAB285276-96
Neomycin ELISA Kit, Inc.
Tests Tests ELISA
A competitive ELISA kit for quantitate of neomycin in tissue, honey, milk, milk powder, liver.
  • Easy, convenient and time-saving method
  • Detection Range: 0,1 to 8,1 ppb
  • Sensitivity: <0,1 ppb
  • Detection limitation: 4 ppb
Neomycin is an aminoglycoside antibiotic found in many topical medications such as creams, ointments, and eyedrops. It is typically used as a topical preparation, such as neosporin. It can also be given orally, where it is usually combined with other antibiotics.
Neomycin, like other aminoglycosides, has been shown to be ototoxic causing tinnitus, hearing loss, and vestibular problems in a small number of patients. Standard techniques/instruments (HPLC or GC-MS) are utilised to detect neomycin.
However, these techniques are complex, expensive, laborious, and time-consuming. Immunoassay techniques, such as ELISAs are commonly preferred as a simple, reliable and rapid method for the quantification of neomycin in various samples.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité