Estrogen Receptor beta (ERβ) (Human) ELISA Kit
Fournisseur: Abcam
AB285292-96
ABCAAB285292-96EA 920 EUR InStock
ABCAAB285292-96
Estrogen Receptor beta (ERβ) (Human) ELISA Kit
Tests Tests ELISA
An Sandwich ELISA kit to quantify human ER-beat in human serum, plasma and other biological fluids.
  • Easy, convenient and time-saving method to measure the level of ER-beta in human serum, plasma, tissue homogenates, culture supernatants and other biological fluids
  • Detection Range: 0,313 to 20 ng/ml
  • Sensitivity: <0,188 ng/ml
Nuclear hormone receptor. Binds estrogens with an affinity similar to that of ESR1, and activates expression of reporter genes containing estrogen response elements (ERE) in an estrogen-dependent manner. Isoform beta-cx lacks ligand binding ability and has no or only very low ere binding activity resulting in the loss of ligand-dependent transactivation ability. DNA-binding by ESR1 and ESR2 is rapidly lost at 37 degrees Celsius in the absence of ligand while in the presence of 17 beta-estradiol and 4-hydroxy-tamoxifen loss in DNA-binding at elevated temperature is more gradual. BioVision’s ER-beta ELISA kit is a sandwich ELISA assay for the quantitative measurement of human ER-beta in serum, plasma and cell culture supernatants. The density of colour is proportional to the amount of human ER-betacaptured from the samples.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité