QuickDetect™ 24(S)-Hydroxycholesterol (Human) ELISA Kit
Fournisseur: Abcam
AB285293-96
ABCAAB285293-96EA 1040 EUR InStock
ABCAAB285293-96
QuickDetect™ 24(S)-Hydroxycholesterol (Human) ELISA Kit
Tests Tests ELISA
A rapid sandwich ELISA kit to quantify CD8 in human serum, plasma, cell culture supernatnat.
  • Easy, convenient and time-saving method to measure the level of 24-OHC in human serum, plasma, culture supernatants and other biological fluids
  • The entire assay can be performed within 90 minutes with only a single incubation and a wash step respectively, resulting in fewer handling steps, fewer errors and more consistent results
  • Detection Range: 0,5 to 100 ng/ml
An important cholesterol metabolite (cholesterol 24-hydroxylase) found in the brain. It is an endogenous agonist of the nuclear receptor LXR. It has been shown to induce cell death in neuroblastoma cells, possibly via a necroptosis pathway. 24(S)-Hydroxycholesterol has also been found to be a high affinity ligand for RORα and RORγ (Ki = 25 nM). 24(S)-Hydroxycholesterol levels are elevated in the cerebral spinal fluids of patients with neurogenerative diseases suggesting possible clinical applications. It was also able to reduce Aβ production and increase ER-resident immature amyloid precursor protein (APP) levels in human neuroblastoma SH-Sy5Y cells and CHO cells via inhibition of intracellular amyloid precursor protein trafficking. BioVision’s QuickDetect™ 24-OHC ELISA kit is a sandwich ELISA assay for measuring the quantity of human 24-OHC in serum, plasma and cell culture and other biological fluid. The entire process can be performed within 90 minutes with only a single incubation and a wash step respectively, resulting in fewer handling steps, fewer errors and more consistent results.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité