Human Native Apolipoprotein B (from Plasma)
Fournisseur: Abcam
AB285810-500
ABCAAB285810-500EA 360 EUR InStock
ABCAAB285810-500
Human Native Apolipoprotein B (from Plasma)
Protéines et Peptides
≥95% pure native Apolipoprotein B isolated from human plasma.
Apolipoprotein B is the dominant protein constituent of LDL. The concentration of Apo B in normal plasma is 90 mg per 100 ml. Apo B is thought to stabilise lipid emulsions, serve as a co-factor and modulator of enzymatic reactions, manage export of lipids out of cells and direct lipids to target organs. Apo B levels are positively correlated with the risk of coronary disease. Apo B levels may be a more sensitive predictor of cardiovascular risk than LDL levels and do not involve fasting for accurate measurement. Two forms of Apo B exist: Apo B-100 and Apo B-48. The first is found in VLDL and LDL and is produced by the liver. The second is found in chylomicrons and originates in the intestine. Mutations in this gene or its regulatory region cause hypobetalipoproteinaemia, normotriglyceridemic and hypercholesterolemia due to ligand-defective Apo B, diseases affecting plasma cholesterol and Apo B levels.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité