96-Well, Black Plates
Fournisseur: Abcam
AB286899-2
ABCAAB286899-2EA 140 EUR InStock
ABCAAB286899-2
96-Well, Black Plates
Microplaques Microplaques pour culture cellulaire
Non-pyrogenic, polystyrene microplates (black) with standard solid flat bottom.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Packaging dimension: 335×250×165 mm
  • The risk of sample carryover and cross-contamination is minimized
  • Suitable for fluorescence measurements
  • The polystyrene surface is more hydrophobic than the surface of a high binding microplate, and therefore tends to be more suitable for non-polar proteins and peptides
These are polystyrene microplates (solid black) with standard solid flat bottom, chimney well. The flat bottom allows precise optics and the rounded corners allow pipetting without leaving a residue.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité