Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

L'expérience en ligne d'Avantor évolue pour mieux vous accompagner ! À compter du 4 avril 2025, nos clients seront migrés vers une nouvelle plateforme pour une expérience d'achat en ligne simplifiée.

QuickDetect™ beta-Endorphin (Human) ELISA Kit

Fournisseur: Abcam

AB287794-96
ABCAAB287794-96EA 979 EUR InStock
ABCAAB287794-96
QuickDetect™ beta-Endorphin (Human) ELISA Kit
Tests Tests ELISA
A rapid ELISA kit to quantify of beta-Endorphin in human serum, plasma, urine, cell culture samples.

  • Easy, convenient and time-saving method to measure the level of beta-Endorphin in human Serum, plasma, urine, cell culture samples, biological fluid
  • Detection Range: 3 to 200 pg/ml
  • Sensitivity: <0,5 pg/ml
  • Cross Reactivity: No significant cross-reactivity or interference between this analyte and its analogues was observed

β-Endorphin (beta-EP) is an endogenous opioid neuropeptide and peptide hormone that is produced in certain neurons within the central nervous system and peripheral nervous system. It is one of five endorphins that are produced in human, the others of which include α-endorphin, γ-endorphin, α-neoendorphin, and β-neoendorphin. Function of β-endorphin has been known to be associated with hunger, thrill, pain, maternal care, sexual behavior, and reward cognition. In the broadest sense, β-endorphin is primarily utilized in the body to reduce stress and maintain homeostasis. In behavioral research, studies have shown that β-endorphin is released via volume transmission into the ventricular system in response to a variety of stimuli, and novel stimuli in particular. BioVision’s beta-EP ELISA kit is a sandwich ELISA assay for the quantitative measurement of beta-EP in human serum, plasma and cell culture supernatants in 90 minutes. The density of colour is proportional to the amount of beta-EP captured from the samples.

Attention:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.




Commander maintenant

 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité

Ce site utilise des cookies, en provenance de VWR ou de ses partenaires, afin de collecter des informations statistiques sur votre navigation et vous proposer des contenus en accord avec vos préférences, générés en fonction de vos habitudes de navigation. En poursuivant la consultation de ce site, vous approuvez l’utilisation de ces cookies.

Pour en savoir plus, consultez notre Politique de Confidentialit&eacute;..