Prostaglandin E2 (PGE2) ELISA Kit
Fournisseur: Abcam
AB287802-96
ABCAAB287802-96EA 0 EUR InStock
ABCAAB287802-96
Prostaglandin E2 (PGE2) ELISA Kit
Tests Tests ELISA
A competitive ELISA kit for the quantitative measurement of prostaglandin E2 in biological fluids.
  • Easy, convenient and time-saving method
  • Detection range: 31,25 to 2000 pg/ml
  • Sensitivity: <18,75 pg/ml Assay Precision: Intra-assay: CV <8%, inter-assay: CV <10%
  • No significant cross-reactivity or interference was observed
Prostaglandin E2 (PGE2) is a primary product of arachidonic metabolism and is synthesized via the cyclooxygenase (COX) and prostaglandin synthase pathways.
PGE2 production is a commonly used method for the detection of COX-1 and COX-2 modulation and prostaglandin synthases.
PGE2 ELISA kit is a competitive ELISA assay for the quantitative measurement of PGE2 in serum, plasma and cell culture supernatants. The density of colour is proportional to the amount of PGE2 captured from the samples.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité