Cortisol ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of cortisol in dried fecal extracts, saliva, urine, serum, plasma and tissue culture media samples 1 * 1 KIT
Fournisseur: Abnova
Cortisol ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of cortisol in dried fecal extracts, saliva, urine, serum, plasma and tissue culture media samples 1 * 1 KIT
COMMANDE
Développer tout / Réduire tout
Cortisol ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of cortisol in dried fecal extracts, saliva, urine, serum, plasma and tissue culture media samples 1 * 1 KIT ABNOKA3382 1 * 1 KIT
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou